Assay Design



Gene Transcripts

Gene Symbol
AAP7
Entrez Gene ID
832446
Gene Name
amino acid permease 7
Gene Aliases
AT5G23810, MRO11.15, MRO11_15, amino acid permease 7
Chromosome Location
Chr.5: 8028238 - 8030888 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036857.1 NP_001031934.1 5 - 6 849 77
NM_001343829.1 NP_001330894.1 4 - 5 1087 77
NM_122286.4 NP_197770.1 5 - 6 989 77

View Full Product Details