Assay Design



Gene Transcripts

Gene Symbol
SQP1
Entrez Gene ID
832480
Gene Name
FAD/NAD(P)-binding oxidoreductase family protein
Gene Aliases
AT5G24150, K12G2.2, K12G2_2, SQE5, SQUALENE MONOOXYGENASE 5
Chromosome Location
Chr.5: 8172300 - 8175525 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036858.1 NP_001031935.1 2 - 3 349 74
NM_122320.4 NP_197803.1 2 - 3 443 74
GenBank mRNA AK316701.1 - 2 - 3 350 74

View Full Product Details