Assay Design



Gene Transcripts

Gene Symbol
PRR5
Entrez Gene ID
832518
Gene Name
two-component response regulator-like protein
Gene Aliases
APRR5, AT5G24470, T31K7.5, T31K7_5, pseudo-response regulator 5
Chromosome Location
Chr.5: 8356204 - 8358546 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122355.4 NP_568446.2 5 - 6 1585 76
GenBank mRNA AY062114.1 - 5 - 6 1705 76
AY143934.1 - 5 - 6 1585 76

View Full Product Details