Assay Design



Gene Transcripts

Gene Symbol
CYP71A14
Entrez Gene ID
832566
Gene Name
cytochrome P450, family 71, subfamily A, polypeptide 14
Gene Aliases
''cytochrome P450, AT5G24960, F6A4.170, F6A4_170, cytochrome P450, family 71, polypeptide 14, polypeptide 14'', subfamily A
Chromosome Location
Chr.5: 8599735 - 8603321 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343905.1 NP_001331829.1 2 - 3 1000 75
NM_122405.2 NP_197878.1 2 - 3 1000 75

View Full Product Details