Assay Design
Gene Transcripts
- Gene Symbol
- DNMT2
- Entrez Gene ID
-
832623
- Gene Name
- DNA methyltransferase-2
- Gene Aliases
-
AT5G25480, AtDNMT2, DNA METHYLTRANSFERASE 2, DNA methyltransferase-2, T14C9.10, T14C9_10
- Chromosome Location
-
Chr.5: 8869865 - 8872262 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122459.3 |
NP_568474.1 |
1 - 2 |
139 |
64 |
View Full Product Details