Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_122490.4 | NP_568478.1 | 1 - 2 | 406 | 94 |
GenBank mRNA | AF149816.1 | - | 1 - 2 | 195 | 94 |
AK228312.1 | - | 1 - 2 | 143 | 94 | |
AY669799.1 | - | 1 - 2 | 130 | 94 | |
BT000427.1 | - | 1 - 2 | 144 | 94 | |
BT002175.1 | - | 1 - 2 | 130 | 94 |