Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_122490.4 | NP_568478.1 | 3 - 4 | 779 | 63 |
GenBank mRNA | AF149816.1 | - | 3 - 4 | 568 | 63 |
AK228312.1 | - | 3 - 4 | 516 | 63 | |
AY669799.1 | - | 3 - 4 | 503 | 63 | |
BT000427.1 | - | 3 - 4 | 517 | 63 | |
BT002175.1 | - | 3 - 4 | 503 | 63 |