Assay Design



Gene Transcripts

Gene Symbol
CLC-D
Entrez Gene ID
832693
Gene Name
chloride channel D
Gene Aliases
AT5G26240, ATCLC-D, AtCLCd, CHLORIDE CHANNEL D, CLCd, T19G15.90, T19G15_90, chloride channel D
Chromosome Location
Chr.5: 9189425 - 9194758 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343969.1 NP_001330038.1 6 - 7 966 71
NM_122525.3 NP_197996.1 6 - 7 981 71

View Full Product Details