Assay Design



Gene Transcripts

Gene Symbol
PWD
Entrez Gene ID
832706
Gene Name
chloroplastidic phosphoglucan, water dikinase (ATGWD3)
Gene Aliases
AT5G26570, ATGWD3, F9D12.1, F9D12_1, OK1, PHOSPHOGLUCAN WATER DIKINASE
Chromosome Location
Chr.5: 9261479 - 9267732 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125808.1 NP_001119280.1 12 - 13 2007 70
NM_122538.5 NP_198009.3 12 - 13 2108 70
GenBank mRNA AJ635427.1 - 12 - 13 2007 70

View Full Product Details