Assay Design



Gene Transcripts

Gene Symbol
ATSAC1
Entrez Gene ID
838868
Gene Name
Phosphoinositide phosphatase family protein
Gene Aliases
AT1G22620, F12K8.3, SAC1, suppressor of actin 1
Chromosome Location
Chr.1: 7994166 - 8002989 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102109.4 NP_173676.2 1 - 2 334 153
GenBank mRNA AF266460.1 - 1 - 2 132 153
AY227244.1 - 1 - 2 132 153

View Full Product Details