Assay Design



Gene Transcripts

Gene Symbol
GI
Entrez Gene ID
838883
Gene Name
gigantea protein (GI)
Gene Aliases
AT1G22770, FB, GIGANTEA, T22J18.6, T22J18_6
Chromosome Location
Chr.1: 8061751 - 8067790 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102124.3 NP_564180.1 8 - 9 1519 67
GenBank mRNA AF105064.1 - 7 - 8 1291 67
AJ133786.1 - 7 - 8 1081 67

View Full Product Details