Assay Design



Gene Transcripts

Gene Symbol
GH9B6
Entrez Gene ID
838930
Gene Name
glycosyl hydrolase 9B6
Gene Aliases
AT1G23210, AtGH9B6, F26F24.6, F26F24_6, glycosyl hydrolase 9B6
Chromosome Location
Chr.1: 8240145 - 8242261 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102170.2 NP_173735.1 1 - 2 646 78
GenBank mRNA BT026379.1 - 1 - 2 617 78
DQ056459.1 - 1 - 2 617 78

View Full Product Details