Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001332600.1 | NP_001320962.1 | 3 - 4 | 516 | 98 |
NM_102191.6 | NP_564194.1 | 3 - 4 | 475 | 98 | |
GenBank mRNA | AB493475.1 | - | 3 - 4 | 390 | 98 |
AF195047.1 | - | 3 - 4 | 417 | 98 | |
BT026410.1 | - | 3 - 4 | 390 | 98 | |
DQ446281.1 | - | 3 - 4 | 390 | 98 |