Assay Design



Gene Transcripts

Gene Symbol
INO
Entrez Gene ID
838950
Gene Name
Plant-specific transcription factor YABBY family protein
Gene Aliases
AT1G23420, F26F24.29, F26F24_29, INNER NO OUTER
Chromosome Location
Chr.1: 8317297 - 8319491 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332600.1 NP_001320962.1 6 - 7 743 71
NM_102191.6 NP_564194.1 6 - 7 702 71
GenBank mRNA AB493475.1 - 6 - 7 617 71
AF195047.1 - 6 - 7 644 71
BT026410.1 - 6 - 7 617 71
DQ446281.1 - 6 - 7 617 71

View Full Product Details