Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001332622.1 | NP_001321290.1 | 9 - 10 | 1502 | 57 |
NM_102228.4 | NP_564204.1 | 10 - 11 | 1410 | 57 | |
GenBank mRNA | AY035139.1 | - | 10 - 11 | 1402 | 57 |
AY113912.1 | - | 10 - 11 | 1328 | 57 |