Assay Design



Gene Transcripts

Gene Symbol
ALDH2B7
Entrez Gene ID
838991
Gene Name
aldehyde dehydrogenase 2B7
Gene Aliases
ALDH2B, AT1G23800, F5O8.35, F5O8_35, aldehyde dehydrogenase 2B, aldehyde dehydrogenase 2B7
Chromosome Location
Chr.1: 8412051 - 8414886 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102228.4 NP_564204.1 8 - 9 1217 87
GenBank mRNA AY035139.1 - 8 - 9 1209 87
AY113912.1 - 8 - 9 1135 87

View Full Product Details