Assay Design



Gene Transcripts

Gene Symbol
SNL3
Entrez Gene ID
839032
Gene Name
SIN3-like 3
Gene Aliases
ARABIDOPSIS THALIANA SIN3 HOMOLOG, AT1G24190, ATSIN3, F3I6.12, F3I6_12, SIN3, SIN3-like 3
Chromosome Location
Chr.1: 8563510 - 8570580 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198151.2 NP_001185080.1 15 - 16 2796 89
NM_001332641.1 NP_001323398.1 15 - 16 2752 89
NM_102265.3 NP_173829.3 15 - 16 2733 89

View Full Product Details