Assay Design



Gene Transcripts

Gene Symbol
OVA9
Entrez Gene ID
839120
Gene Name
glutamine-tRNA ligase, putative / glutaminyl-tRNA synthetase, putative / GlnRS
Gene Aliases
AT1G25350, F4F7.26, F4F7_26, ovule abortion 9
Chromosome Location
Chr.1: 8889018 - 8894306 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198165.1 NP_001185094.1 15 - 16 1600 86
NM_102345.4 NP_173906.2 15 - 16 1630 86
GenBank mRNA AY062594.1 - 15 - 16 1584 86
AY114653.1 - 15 - 16 1529 86

View Full Product Details