Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_102360.4 | NP_173920.1 | 7 - 8 | 1230 | 120 |
GenBank mRNA | AY120757.1 | - | 7 - 8 | 1172 | 120 |
BT000108.1 | - | 6 - 7 | 1055 | 120 | |
U21557.1 | - | 7 - 8 | 1133 | 120 |