Assay Design



Gene Transcripts

Gene Symbol
RCN1
Entrez Gene ID
839135
Gene Name
ARM repeat superfamily protein
Gene Aliases
AT1G25490, ATB BETA BETA, EER1, ENHANCED ETHYLENE RESPONSE 1, F2J7.19, F2J7_19, PHOSPHOPROTEIN PHOSPHATASE 2A REGULATORY SUBUNIT A, REGA, ROOTS CURL IN NPA, SERINE/THREONINE PROTEIN PHOSPHATASE TYPE 2A REGULATORY SUBUNIT A
Chromosome Location
Chr.1: 8951162 - 8955160 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102360.4 NP_173920.1 8 - 9 1334 91
GenBank mRNA AY120757.1 - 8 - 9 1276 91
BT000108.1 - 7 - 8 1159 91
U21557.1 - 8 - 9 1237 91

View Full Product Details