Assay Design



Gene Transcripts

Gene Symbol
CYCT1;3
Entrez Gene ID
839655
Gene Name
cyclin T 1;3
Gene Aliases
AT1G27630, T22C5.8, cyclin T 1;3
Chromosome Location
Chr.1: 9611053 - 9614336 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332773.1 NP_001321314.1 3 - 4 1124 62
NM_001332774.1 NP_001321315.1 2 - 3 1234 62
NM_001332775.1 NP_001321316.1 3 - 4 521 62
NM_001332776.1 NP_001321317.1 3 - 4 903 62
NM_001332777.1 NP_001321318.1 1 - 2 798 62
NM_102528.4 NP_174084.1 3 - 4 571 62
GenBank mRNA AF344323.1 - 2 - 3 315 62
AK117692.1 - 3 - 4 521 62

View Full Product Details