Assay Design
Gene Transcripts
- Gene Symbol
- PUP3
- Entrez Gene ID
-
839716
- Gene Name
- purine permease 3
- Gene Aliases
-
AT1G28220, ATPUP3, F3H9.12, F3H9_12, purine permease 3
- Chromosome Location
-
Chr.1: 9860291 - 9861568 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102587.2 |
NP_174143.1 |
1 - 2 |
823 |
68 |
View Full Product Details