Assay Design
Gene Transcripts
- Gene Symbol
- LEC2
- Entrez Gene ID
-
839724
- Gene Name
- AP2/B3-like transcriptional factor family protein
- Gene Aliases
-
AT1G28300, AtLEC2, F3H9.5, F3H9_5, LEAFY COTYLEDON 2
- Chromosome Location
-
Chr.1: 9896867 - 9899876 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102595.2 |
NP_564304.1 |
3 - 4 |
712 |
132 |
GenBank mRNA |
DQ446296.1 |
- |
3 - 4 |
674 |
132 |
View Full Product Details