Assay Design



Gene Transcripts

Gene Symbol
SFP2
Entrez Gene ID
832795
Gene Name
Major facilitator superfamily protein
Gene Aliases
AT5G27360, F21A20.70, F21A20_70
Chromosome Location
Chr.5: 9656949 - 9662663 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344004.1 NP_001330510.1 13 - 14 1383 82
NM_001344005.1 NP_001330511.1 14 - 15 1230 82
NM_001344006.1 NP_001330508.1 14 - 15 1179 82
NM_001344007.1 NP_001330509.1 11 - 12 1160 82
NM_122618.3 NP_568494.1 14 - 15 1180 82
GenBank mRNA AY026255.1 - 14 - 15 1077 82

View Full Product Details