Assay Design
Gene Transcripts
- Gene Symbol
- LACS7
- Entrez Gene ID
-
832820
- Gene Name
- long-chain acyl-CoA synthetase 7
- Gene Aliases
-
AT5G27600, ATLACS7, F15A18.60, F15A18_60, long-chain acyl-CoA synthetase 7
- Chromosome Location
-
Chr.5: 9742492 - 9747024 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122642.5 |
NP_198112.2 |
6 - 7 |
657 |
91 |
View Full Product Details