Assay Design



Gene Transcripts

Gene Symbol
HTA7
Entrez Gene ID
832829
Gene Name
histone H2A 7
Gene Aliases
AT5G27670, T1G16.3, histone H2A 7
Chromosome Location
Chr.5: 9792509 - 9793560 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122649.5 NP_198119.1 1 - 2 414 90
GenBank mRNA AF385711.1 - 1 - 2 308 90
AY078019.1 - 1 - 2 219 90

View Full Product Details