Assay Design
Gene Transcripts
- Gene Symbol
- BGLU24
- Entrez Gene ID
-
832944
- Gene Name
- beta glucosidase 24
- Gene Aliases
-
AT5G28510, T26D3.6, T26D3_6, beta glucosidase 24
- Chromosome Location
-
Chr.5: 10480757 - 10484073 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122734.2 |
NP_198203.1 |
7 - 8 |
867 |
64 |
View Full Product Details