Assay Design
Gene Transcripts
- Gene Symbol
- SRS8
- Entrez Gene ID
-
833279
- Gene Name
- SHI-related sequence 8
- Gene Aliases
-
AT5G33210, SHI-related sequence 8, T29A4.20, T29A4_20
- Chromosome Location
-
Chr.5: 12464746 - 12465764 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122845.1 |
NP_198306.1 |
1 - 2 |
96 |
67 |
View Full Product Details