Assay Design



Gene Transcripts

Gene Symbol
EGY1
Entrez Gene ID
833476
Gene Name
Peptidase M50 family protein
Gene Aliases
AMOS1, AT5G35220, ETHYLENE-DEPENDENT GRAVITROPISM-DEFICIENT AND YELLOW-GREEN 1, T25C13.100, T25C13_100, ammonium overly sensitive 1
Chromosome Location
Chr.5: 13484403 - 13487594 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122913.4 NP_198372.1 2 - 3 837 93
GenBank mRNA AY050809.1 - 2 - 3 790 93
BT001978.1 - 2 - 3 789 93

View Full Product Details