Assay Design
Gene Transcripts
- Gene Symbol
- HK2
- Entrez Gene ID
-
833552
- Gene Name
- histidine kinase 2
- Gene Aliases
-
AHK2, AT5G35750, MXH1.16, MXH1_16, histidine kinase 2
- Chromosome Location
-
Chr.5: 13911283 - 13916805 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122966.3 |
NP_568532.1 |
3 - 4 |
1393 |
151 |
GenBank mRNA |
AB046869.1 |
- |
3 - 4 |
925 |
151 |
View Full Product Details