Assay Design



Gene Transcripts

Gene Symbol
CYB-1
Entrez Gene ID
833853
Gene Name
cytochrome B561-1
Gene Aliases
ACYB-1, AT5G38630, MBB18.18, MBB18_18, cytochrome B561-1
Chromosome Location
Chr.5: 15465992 - 15468185 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123224.3 NP_198679.1 2 - 3 340 145
GenBank mRNA AB049627.1 - 2 - 3 316 145
AK117270.1 - 2 - 3 299 145
AY085028.1 - 2 - 3 340 145
BT024611.1 - 2 - 3 191 145

View Full Product Details