Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_123224.3 | NP_198679.1 | 3 - 4 | 546 | 66 |
GenBank mRNA | AB049627.1 | - | 3 - 4 | 522 | 66 |
AK117270.1 | - | 3 - 4 | 505 | 66 | |
AY085028.1 | - | 3 - 4 | 546 | 66 | |
BT024611.1 | - | 3 - 4 | 397 | 66 |