Assay Design
Gene Transcripts
- Gene Symbol
- cdc2cAt
- Entrez Gene ID
-
833938
- Gene Name
- CDC2C
- Gene Aliases
-
AT5G39420, CDC2C, MUL8.100, MUL8_100
- Chromosome Location
-
Chr.5: 15771931 - 15775251 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123304.4 |
NP_198758.2 |
6 - 7 |
1671 |
112 |
GenBank mRNA |
BT046183.1 |
- |
6 - 7 |
1370 |
112 |
View Full Product Details