Assay Design
Gene Transcripts
- Gene Symbol
- GLA1
- Entrez Gene ID
-
834149
- Gene Name
- Folylpolyglutamate synthetase family protein
- Gene Aliases
-
A. THALIANA DHFS-FPGS HOMOLOG A, AT5G41480, ATDFA, DFA, DHFS-FPGS HOMOLOG A, EMB9, EMBRYO DEFECTIVE 9, GLOBULAR ARREST1, MYC6.18, MYC6_18
- Chromosome Location
-
Chr.5: 16595863 - 16598682 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123512.3 |
NP_198963.2 |
3 - 4 |
637 |
129 |
View Full Product Details