Assay Design



Gene Transcripts

Gene Symbol
scpl41
Entrez Gene ID
834228
Gene Name
serine carboxypeptidase-like 41
Gene Aliases
AT5G42230, K5J14.3, K5J14_3, serine carboxypeptidase-like 41
Chromosome Location
Chr.5: 16880592 - 16884462 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344440.1 NP_001318730.1 5 - 6 775 75
NM_123588.4 NP_199038.1 4 - 5 935 75
GenBank mRNA BT002948.1 - 4 - 5 568 75
BT004386.1 - 3 - 4 437 75

View Full Product Details