Assay Design



Gene Transcripts

Gene Symbol
scpl42
Entrez Gene ID
834229
Gene Name
serine carboxypeptidase-like 42
Gene Aliases
AT5G42240, K5J14.4, K5J14_4, serine carboxypeptidase-like 42
Chromosome Location
Chr.5: 16887626 - 16891192 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123589.4 NP_199039.1 6 - 7 1303 98
GenBank mRNA AF361604.1 - 6 - 7 696 98
AY143899.1 - 5 - 6 625 98

View Full Product Details