Assay Design
Gene Transcripts
- Gene Symbol
- DFR
- Entrez Gene ID
-
834291
- Gene Name
- dihydroflavonol 4-reductase
- Gene Aliases
-
AT5G42800, DIHYDROFLAVONOL 4-REDUCTASE, DIHYDROKAEMPFEROL 4-REDUCTASE, M318, MJB21.18, MJB21_18, TT3, dihydroflavonol 4-reductase
- Chromosome Location
-
Chr.5: 17164093 - 17166032 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123645.4 |
NP_199094.1 |
2 - 3 |
456 |
69 |
View Full Product Details