Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_123663.3 | NP_199111.1 | 5 - 6 | 699 | 150 |
GenBank mRNA | AF395060.1 | - | 5 - 6 | 532 | 150 |
AK176360.1 | - | 5 - 6 | 614 | 150 | |
BT025548.1 | - | 5 - 6 | 532 | 150 |