Assay Design



Gene Transcripts

Gene Symbol
ckl8
Entrez Gene ID
834350
Gene Name
casein kinase I-like 8
Gene Aliases
AT5G43320, MWF20.1, MWF20_1, casein kinase I-like 8
Chromosome Location
Chr.5: 17385760 - 17389600 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344480.1 NP_001330706.1 11 - 12 1584 62
NM_001344481.1 NP_001330707.1 11 - 12 1534 62
NM_123698.4 NP_199146.1 11 - 12 1584 62
GenBank mRNA AF360257.1 - 11 - 12 1334 62
AY040071.1 - 11 - 12 925 62
AY943854.1 - 11 - 12 925 62

View Full Product Details