Assay Design
Gene Transcripts
- Gene Symbol
- PDLP1
- Entrez Gene ID
-
834421
- Gene Name
- plasmodesmata-located protein 1
- Gene Aliases
-
AT5G43980, MRH10.9, MRH10_9, PDLP1A, PLASMODESMATA-LOCATED PROTEIN 1A, plasmodesmata-located protein 1
- Chromosome Location
-
Chr.5: 17694680 - 17696690 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123765.3 |
NP_199211.2 |
1 - 2 |
1004 |
104 |
View Full Product Details