Assay Design
Gene Transcripts
- Gene Symbol
- RTL3
- Entrez Gene ID
-
834551
- Gene Name
- RNAse THREE-like protein 3
- Gene Aliases
-
AT5G45150, K18C1.3, K18C1_3, RNAse THREE-like protein 3
- Chromosome Location
-
Chr.5: 18260313 - 18264454 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123883.2 |
NP_199328.1 |
3 - 4 |
605 |
99 |
View Full Product Details