Assay Design



Gene Transcripts

Gene Symbol
RTL3
Entrez Gene ID
834551
Gene Name
RNAse THREE-like protein 3
Gene Aliases
AT5G45150, K18C1.3, K18C1_3, RNAse THREE-like protein 3
Chromosome Location
Chr.5: 18260313 - 18264454 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344592.1 NP_001330579.1 4 - 5 1772 108
NM_001344593.1 NP_001330578.1 4 - 5 1697 108
NM_123883.2 NP_199328.1 5 - 6 1534 108

View Full Product Details