Assay Design
Gene Transcripts
- Gene Symbol
- RLP55
- Entrez Gene ID
-
834616
- Gene Name
- receptor like protein 55
- Gene Aliases
-
AT5G45770, AtRLP55, MRA19.20, MRA19_20, receptor like protein 55
- Chromosome Location
-
Chr.5: 18563333 - 18564921 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123944.3 |
NP_199389.1 |
1 - 1 |
1522 |
76 |
View Full Product Details