Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001344708.1 | NP_001330804.1 | 3 - 4 | 491 | 138 |
NM_124081.4 | NP_199521.1 | 3 - 4 | 512 | 138 | |
GenBank mRNA | AB060590.1 | - | 2 - 3 | 166 | 138 |
AF411958.1 | - | 2 - 3 | 166 | 138 | |
BT002995.1 | - | 3 - 4 | 344 | 138 | |
BT004458.1 | - | 2 - 3 | 166 | 138 |