Assay Design



Gene Transcripts

Gene Symbol
TDT
Entrez Gene ID
834806
Gene Name
tonoplast dicarboxylate transporter
Gene Aliases
AT5G47560, ATSDAT, ATTDT, TONOPLAST DICARBOXYLATE TRANSPORTER, tonoplast dicarboxylate transporter
Chromosome Location
Chr.5: 19287369 - 19291103 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124129.4 NP_199567.1 1 - 2 1200 67
GenBank mRNA AB043024.1 - 1 - 2 477 67
AY072145.1 - 1 - 2 495 67
AY133792.1 - 1 - 2 444 67

View Full Product Details