Assay Design



Gene Transcripts

Gene Symbol
OVA2
Entrez Gene ID
834962
Gene Name
tRNA synthetase class I (I, L, M and V) family protein
Gene Aliases
AT5G49030, K19E20.18, K19E20_18, ovule abortion 2
Chromosome Location
Chr.5: 19874925 - 19883297 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036965.2 NP_001032042.1 6 - 7 894 103
NM_001203568.1 NP_001190497.1 7 - 8 1327 103
NM_124280.4 NP_199714.2 6 - 7 1006 103
GenBank mRNA AY080830.1 - 6 - 7 823 103

View Full Product Details