Assay Design



Gene Transcripts

Gene Symbol
MET1
Entrez Gene ID
834975
Gene Name
methyltransferase 1
Gene Aliases
AT5G49160, DDM2, DECREASED DNA METHYLATION 2, DMT01, DMT1, DNA METHYLTRANSFERASE, DNA METHYLTRANSFERASE 01, DNA METHYLTRANSFERASE 1, MET2, METHYLTRANSFERASE 2, METHYLTRANSFERASE I, METI, methyltransferase 1
Chromosome Location
Chr.5: 19932114 - 19938470 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344823.1 NP_001331134.1 8 - 9 3461 134
NM_124293.4 NP_199727.1 7 - 8 3770 134
GenBank mRNA L10692.1 - 7 - 8 3663 134

View Full Product Details