Assay Design



Gene Transcripts

Gene Symbol
PAP27
Entrez Gene ID
835108
Gene Name
purple acid phosphatase 27
Gene Aliases
ARABIDOPSIS THALIANA PURPLE ACID PHOSPHATASE 27, AT5G50400, ATPAP27, MXI22.12, MXI22_12, purple acid phosphatase 27
Chromosome Location
Chr.5: 20523366 - 20526521 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344892.1 NP_001330206.1 4 - 5 664 103
NM_124422.4 NP_199851.2 4 - 5 739 103
GenBank mRNA AY842027.1 - 4 - 5 449 103

View Full Product Details