Assay Design
Gene Transcripts
- Gene Symbol
- BOB1
- Entrez Gene ID
-
835421
- Gene Name
- HSP20-like chaperones superfamily protein
- Gene Aliases
-
AT5G53400, BOBBER1, MYN8.1, MYN8_1
- Chromosome Location
-
Chr.5: 21661444 - 21663843 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124719.4 |
NP_200152.1 |
5 - 6 |
1019 |
95 |
GenBank mRNA |
AK227305.1 |
- |
5 - 6 |
960 |
95 |
View Full Product Details