Assay Design



Gene Transcripts

Gene Symbol
CPK25
Entrez Gene ID
818162
Gene Name
calcium-dependent protein kinase 25
Gene Aliases
AT2G35890, F11F19.20, F11F19_20, calcium-dependent protein kinase 25
Chromosome Location
Chr.2: 15066614 - 15069359 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336578.1 NP_001318361.1 5 - 6 1666 132
NM_129148.2 NP_181133.1 5 - 6 1666 132
GenBank mRNA DQ446601.1 - 5 - 6 1443 132

View Full Product Details